Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-IgG Goat Antibody
Anti-IgG Goat Antibody
Catalog # 490016-834
Supplier:  Advansta
undefined
Anti-IgG Goat Antibody
Catalog # 490016-834
Supplier:  Advansta
Supplier Number:  R-05058-250

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Host:
    Goat
  • Isotype:
    IgG
  • Size:
    250 µL
  • Cat. No.:
    490016-834
  • Supplier no.:
    R-05058-250

Specifications

About this item

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: