Anti-IgM Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Supplier: Columbia Biosciences
Certificates
About this item
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgM
Reactivity: Human
Specifications
- Catalog No:
- 10065-834
- Antigen Symbol:
- IgM
- Antigen Name:
- Immunoglobulin M
- Antibody Type:
- Secondary
- Clonality:
- Polyclonal
- Conjugation:
- RPE (R-Phycoerythrin)
- Reactivity:
- Human
- Host:
- Goat
- Isotype:
- IgM
- ELISA:
- Yes
- Flow Cytometry:
- Yes
- Storage Temperature:
- Store at 4 °C
- Tested Applications:
- Cell-Based Assays, Microarrays and Microplate Applications
Specifications
Frequently Bought Together