Anti-IgE Goat Polyclonal Antibody (APC (Allophycocyanin))
Catalog # 10065-778
Supplier: Columbia Biosciences
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin E
- Antigen Symbol:IgE
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- Isotype:IgE
- Reactivity:Human
- Storage Temperature:Store at 4 °C
- Tested Applications:Cell-Based Assays, Microarrays and Microplate Applications
- Cat. No.:10065-778
- Supplier no.:D3-110-E
Specifications
About this item
Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgE
Reactivity: Human