Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # 10065-886
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # 10065-886
Supplier Number:  D5-112-FABFC

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    RPE (R-Phycoerythrin)
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Storage Temperature:
    Store at 4 °C
  • Tested Applications:
    Cell-Based Assays, Microarrays and Microplate Applications
  • Cat. No.:
    10065-886
  • Supplier no.:
    D5-112-FABFC

Specifications

About this item

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse