Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-IgG1 Goat Polyclonal Antibody (APC (Allophycocyanin))
Anti-IgG1 Goat Polyclonal Antibody (APC (Allophycocyanin))
Catalog # 10065-586
Anti-IgG1 Goat Polyclonal Antibody (APC (Allophycocyanin))
Catalog # 10065-586
Supplier Number:  D3-112-1

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G1
  • Antigen Symbol:
    IgG1
  • Clonality:
    Polyclonal
  • Conjugation:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG1
  • Reactivity:
    Mouse
  • Storage Temperature:
    Store at 4 °C
  • Tested Applications:
    High Throughput Screening, Fluorescence Resonance Energy Transfer Assays, Flow Cytometry and Microscopy
  • Cat. No.:
    10065-586
  • Supplier no.:
    D3-112-1

Specifications

About this item

Type: Secondary
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG1
Reactivity: Mouse