Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # 10065-892
Supplier: Columbia Biosciences
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:RPE (R-Phycoerythrin)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- Reactivity:Rat
- Epitope:F(c)
- Storage Temperature:Store at 4 °C
- Tested Applications:Cell-Based Assays, Microarrays and Microplate Applications
- Cat. No.:10065-892
- Supplier no.:D5-113-FC
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: F(c)
Host: Goat
Isotype:
Reactivity: Rat