Anti-IgG Polyclonal Antibody (SureLight® P3)
Catalog # 10159-224
Supplier: Columbia Biosciences
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:SureLight® P3
- Flow Cytometry:Yes
- Isotype:IgG
- Storage Temperature:Store at 4 °C
- Tested Applications:Cell-Based Assays, Microarrays and Microplate Applications
- Cat. No.:10159-224
- Supplier no.:D7-110-1MG
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: SureLight® P3
Epitope:
Host:
Isotype: IgG
Reactivity: