Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Catalog # 10065-608
Supplier: Columbia Biosciences
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat
- Storage Temperature:Store at 4 °C
- Tested Applications:High Throughput Screening, Fluorescence Resonance Energy Transfer Assays, Flow Cytometry and Microscopy
- Cat. No.:10065-608
- Supplier no.:D3-113
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat