Anti-IgG Polyclonal Antibody (DyLight® 650)
Catalog # 10159-602
Supplier: Columbia Biosciences
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Flow Cytometry:Yes
- Isotype:IgG
- Storage Temperature:Store at 4 °C
- Tested Applications:Cell-Based Assays
- Cat. No.:10159-602
- Supplier no.:D10-114-1MG
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host:
Isotype: IgG
Reactivity: