Anti-V5 Antibody (Europium 1024)
Supplier: Columbia Biosciences
Certificates
About this item
Type:
Antigen: V5
Clonality: 0
Clone:
Conjugation: Europium 1024
Epitope:
Host:
Isotype: IgG
Reactivity:
Specifications
- Catalog No:
- 10159-248
- 10159-250
- 10159-246
- Antigen Symbol:
- V5
- V5
- V5
- Conjugation:
- Europium 1024
- Europium 1024
- Europium 1024
- Isotype:
- IgG
- IgG
- IgG
- Storage Temperature:
- Store at 4 °C
- Store at 4 °C
- Store at 4 °C
- Tested Applications:
- Optimized for TR-FRET High Throughput Screening (HTS) Applications (LANCE®, HTRF®)
- Optimized for TR-FRET High Throughput Screening (HTS) Applications (LANCE®, HTRF®)
- Optimized for TR-FRET High Throughput Screening (HTS) Applications (LANCE®, HTRF®)