Anti-GKPIPNPLLGLDST Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # 10065-742
Supplier: Columbia Biosciences
Specifications
- Antibody Type:Tag
- Antigen Symbol:GKPIPNPLLGLDST
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Storage Temperature:Store at 4 °C
- Tested Applications:High Throughput Screening, Fluorescence Resonance Energy Transfer Assays Targeting Kinase Activity and Flow Cytometry
- Cat. No.:10065-742
- Supplier no.:D9-1834
Specifications
About this item
Type:
Antigen: GKPIPNPLLGLDST
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: