Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MASMTGGQQMG Rabbit Polyclonal Antibody (DyLight® 550)
Anti-MASMTGGQQMG Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # 10065-740
Anti-MASMTGGQQMG Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # 10065-740
Supplier Number:  D9-1832

Specifications

  • Antigen Symbol:
    MASMTGGQQMG
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 550
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Storage Temperature:
    Store at 4 °C
  • Tested Applications:
    High Throughput Screening, Fluorescence Resonance Energy Transfer Assays Targeting Kinase Activity and Flow Cytometry
  • Cat. No.:
    10065-740
  • Supplier no.:
    D9-1832

Specifications

About this item

Type:
Antigen: MASMTGGQQMG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: