Anti-MASMTGGQQMG Rabbit Polyclonal Antibody (Biotin)
Catalog # 10065-470
Supplier: Columbia Biosciences
Specifications
- Antigen Symbol:MASMTGGQQMG
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Storage Temperature:Store at 4 °C
- Tested Applications:High Throughput Screening
- Cat. No.:10065-470
- Supplier no.:B1-1832
Specifications
About this item
Type:
Antigen: MASMTGGQQMG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: