Anti-KETAAAKFERQHMDS Rabbit Polyclonal Antibody (DyLight® 650)
10065-502
- Antibody Type:Tag
- Antigen Name:KETAAAKFERQHMDS
- Antigen Symbol:KETAAAKFERQHMDS
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Storage Temperature:Store at 4 °C
- Tested Applications:Cell-Based Assays
- Cat. No.:10065-502
- Supplier no.:D10-1831
Type:
Antigen: KETAAAKFERQHMDS
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: