Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-HA Monoclonal Antibody (Europium 1024) [clone: 16B12]
Anti-HA Monoclonal Antibody (Europium 1024) [clone: 16B12]
Certificates
Anti-HA Monoclonal Antibody (Europium 1024) [clone: 16B12]

Specifications

  • Catalog No:
  • 10159-240
  • 10159-242
  • 10159-244
  • Antigen Symbol:
  • HA
  • HA
  • HA
  • Antigen Name:
  • Hemagglutinin
  • Hemagglutinin
  • Hemagglutinin
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Europium 1024
  • Europium 1024
  • Europium 1024
  • Clone:
  • 16B12
  • 16B12
  • 16B12
  • Isotype:
  • IgG
  • IgG
  • IgG
  • Storage Temperature:
  • Store at 4 °C
  • Store at 4 °C
  • Store at 4 °C
  • Tested Applications:
  • Optimized for TR-FRET High Throughput Screening (HTS) Applications (LANCE®, HTRF®)
  • Optimized for TR-FRET High Throughput Screening (HTS) Applications (LANCE®, HTRF®)
  • Optimized for TR-FRET High Throughput Screening (HTS) Applications (LANCE®, HTRF®)

Specifications

Related Information