Anti-HA Monoclonal Antibody (DyLight® 650) [clone: 16B12]
Catalog # 10065-490
Supplier: Columbia Biosciences
Specifications
- Antibody Type:Tag
- Antigen Name:Hemagglutinin
- Antigen Symbol:HA
- Clonality:Monoclonal
- Clone:16B12
- Conjugation:DyLight® 650
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Size:100 µg
- Form:liquid
- Storage Buffer:pH 8.2
- Storage Temperature:Store at 4 °C
- Shipping Temperature:4 °C
- Immunogen:synthetic peptide YPYDVPDYA corresponding to HA tag
- Tested Applications:Cell-Based Assays
- Purification:protein G purified
- Cat. No.:10065-490
- Packaging:Plastic vial
- Supplier no.:D10-1722-100
Specifications
About this item
Type:
Antigen: HA
Clonality: Monoclonal
Clone: 16B12
Conjugation: DyLight® 650
Epitope:
Host:
Isotype: IgG
Reactivity: