Anti-DYKDDDDK Mouse Polyclonal Antibody (DyLight® 550) [clone: M2]
Catalog # 10065-732
Supplier: Columbia Biosciences
Specifications
- Antigen Symbol:DYKDDDDK
- Clonality:Polyclonal
- Clone:M2
- Conjugation:DyLight® 550
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Storage Temperature:Store at 4 °C
- Tested Applications:High Throughput Screening, Fluorescence Resonance Energy Transfer Assays Targeting Kinase Activity and Flow Cytometry
- Cat. No.:10065-732
- Supplier no.:D9-1718
Specifications
About this item
Type:
Antigen: DYKDDDDK
Clonality: Polyclonal
Clone: M2
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: