Anti-DYKDDDDK Mouse Monoclonal Antibody [clone: M2] (APC (Allophycocyanin))
Supplier: Columbia Biosciences
Certificates
About this item
Mouse monoclonal [M2] to DDDDK tag (binds to FLAG sequence) (SureLight Allophycocyanin)
Type:
Antigen: DYKDDDDK
Clonality: Monoclonal
Clone: M2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Specifications
- Catalog No:
- 10065-794
- 10065-792
- Antigen Symbol:
- DYKDDDDK
- DYKDDDDK
- Antibody Type:
- Tag
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- M2
- M2
- Host:
- Mouse
- Mouse
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- synthetic peptide corresponds to DDDDK tag (binds to FLAG tag sequence)
- synthetic peptide corresponds to DDDDK tag (binds to FLAG tag sequence)
- Form:
- liquid
- liquid
- Concentration:
- 100 µg at 0.1mg/ml
- 100 µg at 0.1mg/ml
- Storage Buffer:
- pH 7.4
- pH 7.4
- Storage Temperature:
- Store at 4 °C
- Store at 4 °C
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- FRET
- FRET