Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-6xHis Mouse Monoclonal Antibody [clone: AD1.1.10] (HRP (Horseradish Peroxidase))
Anti-6xHis Mouse Monoclonal Antibody [clone: AD1.1.10] (HRP (Horseradish Peroxidase))
Catalog # 10065-762
Anti-6xHis Mouse Monoclonal Antibody [clone: AD1.1.10] (HRP (Horseradish Peroxidase))
Catalog # 10065-762
Supplier Number:  HRP-1711

Specifications

  • Antibody Type:
    Tag
  • Antigen Name:
    6X HIS Affinity Tag
  • Antigen Symbol:
    6xHis
  • Clonality:
    Monoclonal
  • Clone:
    AD1.1.10
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Form:
    lyophilized
  • Antigen Synonyms:
    Histidine Epitope Tag|His Tag|Histidine Tag
  • Storage Buffer:
    pH 7.4
  • Storage Temperature:
    Store at 4 °C
  • Shipping Temperature:
    4 °C
  • Purification:
    size exclusion
  • Cat. No.:
    10065-762
  • Packaging:
    Plastic vial
  • Supplier no.:
    HRP-1711

Specifications

About this item

Mouse Monocolonal [AD1.1.10] to 6X His tag (HRP)

Type:
Antigen: 6xHis
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: