Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-6xHis Mouse Monoclonal Antibody (DyLight® 650) [clone: AD1.1.1]
Anti-6xHis Mouse Monoclonal Antibody (DyLight® 650) [clone: AD1.1.1]
Certificates
Anti-6xHis Mouse Monoclonal Antibody (DyLight® 650) [clone: AD1.1.1]

Specifications

  • Catalog No:
  • 10065-478
  • 10065-480
  • Antigen Symbol:
  • 6xHis
  • 6xHis
  • Antigen Name:
  • 6X HIS Affinity Tag
  • 6X HIS Affinity Tag
  • Antigen Synonyms:
  • Histidine Epitope Tag|His Tag|Histidine Tag|HHHHHH
  • Histidine Epitope Tag|His Tag|Histidine Tag|HHHHHH
  • Antibody Type:
  • Tag
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • DyLight® 650
  • DyLight® 650
  • Clone:
  • AD1.1.10
  • AD1.1.10
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • synthetic peptide corresponding to 6X His tag
  • synthetic peptide corresponding to 6X His tag
  • Form:
  • liquid
  • liquid
  • Purification:
  • protein G purified
  • protein G purified
  • Concentration:
  • 100 µg at 1mg/ml
  • 100 µg at 1mg/ml
  • Storage Buffer:
  • pH 8.2
  • pH 8.2
  • Storage Temperature:
  • Store at 4 °C
  • Store at 4 °C
  • Shipping Temperature:
  • 4 °C
  • 4 °C
  • Tested Applications:
  • Cell-Based Assays
  • High Throughput Screening, Flow Cytometry, Cell-Based Assays, Microarrays and Microplate Applications

Specifications

Related Information