Anti-EGFR Mouse Monoclonal Antibody [clone: AY13]
Purified anti-human EGFR [AY13]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IF; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For immunofluorescence microscopy, the suggested use of this reagent is 1-5 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: AY13
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
- Catalog No:
- 352902-BL
- 352901-BL
- Antigen Symbol:
- EGFR
- EGFR
- Antigen Synonyms:
- HER1|proto-oncogene c-ErbB-1|Receptor tyrosine-protein kinase erbB-1|Epidermal growth factor receptor
- HER1|proto-oncogene c-ErbB-1|Receptor tyrosine-protein kinase erbB-1|Epidermal growth factor receptor
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- AY13
- AY13
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 1956
- 1956
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Non-small cell lung cancer (NSCLC) cell line NCI-H322
- Non-small cell lung cancer (NSCLC) cell line NCI-H322
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Shipping Temperature:
- Ambient RT
- Ambient RT