Anti-CD156c (ADAM10) Mouse Monoclonal Antibody [clone: SHM14]
352702-BL
- Antibody Type:Primary
- Antigen Symbol:CD156c (ADAM10)
- Clonality:Monoclonal
- Clone:SHM14
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Baboon,African green monkey,Cynomolgus monkey,Rhesus monkey,Human
- Size:100 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:4684
- Antigen Synonyms:ADAM10|MADM|CD156C|KUZ
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:Human A375 cell lines
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:352702-BL
- Supplier no.:352702
Purified anti-human CD156c (ADAM10) [SHM14]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD156c (ADAM10)
Clonality: Monoclonal
Clone: SHM14
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus