Anti-TIGIT (Vstm3) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1G9]
Supplier: Biolegend
Certificates
About this item
PE anti-mouse TIGIT (Vstm3) [1G9]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: TIGIT (Vstm3)
Clonality: Monoclonal
Clone: 1G9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Specifications
- Catalog No:
- 142104-BL
- 142103-BL
- Antigen Symbol:
- TIGIT (Vstm3)
- TIGIT (Vstm3)
- Antigen Synonyms:
- T cell immunoreceptor with Ig and ITIM domains|TIGIT|Vstm3|V-set and transmembrane domain-containing protein 3|VSIG9
- T cell immunoreceptor with Ig and ITIM domains|TIGIT|Vstm3|V-set and transmembrane domain-containing protein 3|VSIG9
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- 1G9
- 1G9
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 100043314
- 100043314
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow Cytometry:
- Yes
- Yes
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT