About this item
PE anti-mouse IL-12/IL-23 p40 [C15.6]; Isotype: Rat IgG1, κ; Reactivity: Mouse, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ICFC; Size: 100 μg
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: IL-12 p40
Clonality: Monoclonal
Clone: C15.6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse;IL-12/IL-23 p40 subunit (monomer;homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40)
Specifications
- Catalog No:
- 505204-BL
- 505203-BL
- Antigen Symbol:
- IL-12 p40
- IL-12 p40
- Antigen Synonyms:
- IL12 + IL23|TcMF p40|NKSF p40|TSF p40|CLMF p40|Interleukin-12 p40|Interleukin-23 p40
- IL12 + IL23|TcMF p40|NKSF p40|TSF p40|CLMF p40|Interleukin-12 p40|Interleukin-23 p40
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- C15.6
- C15.6
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 16160
- 16160
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- CHO-expressed, recombinant mouse IL-12 p70
- CHO-expressed, recombinant mouse IL-12 p70
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT