Anti-CD206 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: C068C2]
FITC anti-mouse CD206 (MMR) [C068C2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, FC; Size: 200 μg
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD206
Clonality: Monoclonal
Clone: C068C2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
- Catalog No:
- 141704-BL
- 141703-BL
- Antigen Symbol:
- CD206
- CD206
- Antigen Synonyms:
- CD206|MMR (macrophage mannose receptor)|MRC1|MR (mannose receptor)
- CD206|MMR (macrophage mannose receptor)|MRC1|MR (mannose receptor)
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- C068C2
- C068C2
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 17533
- 17533
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Recombinant mouse CD206 (MMR)
- Recombinant mouse CD206 (MMR)
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT