To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD90.1
- Clonality:Monoclonal
- Clone:OX-7
- Conjugation:Brilliant Violet® 421
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Rabbit,Guinea pig,Rat,Mouse
- Size:125 μl
- Environmentally Preferable:
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Gene ID:21838, 24832
- Antigen Synonyms:Thy-1|Thy-1.1|THY1
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:Ambient RT
- Immunogen:Rat thymocyte Thy-1 antigen
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 421™ and unconjugated antibody.
- Cat. No.:202529-BL
- Supplier no.:202529
Specifications
About this item
Brilliant Violet 421™ anti-rat CD90/mouse CD90.1 (Thy-1.1) [OX-7]; Isotype: Mouse IgG1, κ; Reactivity: Rat, Mouse (AKR/J and PL mouse strains). Cross-Reactivity: Rabbit (Lapine), Guinea Pig6; Apps: FC; Size: 125 μl
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD90.1
Clonality: Monoclonal
Clone: OX-7
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Rat;Mouse (AKR/J and PL mouse strains). Cross-Reactivity: Rabbit (Lapine);Guinea Pig6