Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD8 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SK1]
Anti-CD8 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SK1]
Supplier:  Biolegend
Certificates
Anti-CD8 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SK1]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 344722-BL
  • 344721-BL
  • Antigen Symbol:
  • CD8
  • CD8
  • Antigen Synonyms:
  • T8|Leu2
  • T8|Leu2
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • SK1
  • SK1
  • Reactivity:
  • African green monkey, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human
  • African green monkey, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 925
  • 925
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
  • The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications