Anti-PPIL2 Rabbit Polyclonal Antibody
89516-924
: Abgent
- Antibody Type:Primary
- Antigen Name:Peptidylprolyl Isomerase (cyclophilin)-like 2
- Antigen Symbol:PPIL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Gene ID:23759
- Antigen Synonyms:CYC4|CYP60|UBOX7|Cyp-60|hCyP-60
- Amino Acid Number:402 to 431
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:58.823 kDa
- Sequence:PKTDRPKEEIRIDATC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This PPIL2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 402-431 amino acids from the C-terminal region of human PPIL2.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89516-924
- Supplier no.:AP18215B
- Western Blot: 1:1000
Type: Primary
Antigen: PPIL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human