Anti-CD317 (BST2, PDCA-1) Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: 129c1]
Alexa Fluor® 647 anti-mouse CD317 (BST2, PDCA-1) [129c1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
* Alexa Fluor® 647 has a maximum emission of 668 nm when it is excited at 633 nm / 635 nm.
Type: Primary
Antigen: CD317 (BST2, PDCA-1)
Clonality: Monoclonal
Clone: 129c1
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
- Catalog No:
- 127106-BL
- 127105-BL
- Antigen Symbol:
- CD317 (BST2, PDCA-1)
- CD317 (BST2, PDCA-1)
- Antigen Synonyms:
- CD317|plasmacytoid DC|BST2|plasmacytoid DC (pDC) Ag-1|PDCA-1
- CD317|plasmacytoid DC|BST2|plasmacytoid DC (pDC) Ag-1|PDCA-1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 647
- Alexa Fluor® 647
- Clone:
- 129c1
- 129c1
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 69550
- 69550
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Mouse plasmacytoid dendritic cells (DCs)
- Mouse plasmacytoid dendritic cells (DCs)
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
- The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Frequently Bought Together