Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 82022-744
Supplier:  G-Biosciences
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 82022-744
Supplier:  G-Biosciences
Supplier Number:  786-R39

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Rabbit
  • Western Blot:
    Yes
  • Size:
    2 mL
  • Cat. No.:
    82022-744
  • Supplier no.:
    786-R39

Specifications

About this item

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit