To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:IgG (minimal x-reactivity)
- Clonality:Polyclonal
- Clone:Poly4053
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Gel Shift:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:500 μg
- Environmentally Preferable:
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:16059
- Antigen Synonyms:IgG
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:405301-BL
- Supplier no.:405301
Specifications
About this item
Purified Goat anti-mouse IgG (minimal x-reactivity) [Poly4053]; Isotype: Goat Polyclonal IgG; Reactivity: Mouse; Apps: ELISA, FC, WB, IHC; Size: 500 μg
Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture, the suggested use of this reagent is 2.0 μg/ml. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: IgG (minimal x-reactivity)
Clonality: Polyclonal
Clone: Poly4053
Conjugation:
Epitope:
Host: Goat
Isotype: Goat Polyclonal IgG
Reactivity: Mouse