Anti-AMPD3 Rabbit Polyclonal Antibody
89521-818
: Abgent
- Antibody Type:Primary
- Antigen Name:AMP deaminase 3
- Antigen Symbol:AMPD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 µL
- Epitope:Center
- Gene ID:272
- Antigen Synonyms:AMP deaminase 3|Erythrocyte AMP deaminase|AMPD3|AMP deaminase isoform E
- Amino Acid Number:325 to 356
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:88.812 kDa
- Sequence:EPDRTVAEKRGRKITLRC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This AMPD3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 325-356 amino acids from the Central region of human AMPD3.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89521-818
- Supplier no.:AP7869C
Western Blot: 1:1000
Type: Primary
Antigen: AMPD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse