About this item
Purified anti-human IL-12/IL-23 p40 [C11.5]; Isotype: Mouse IgG1, κ; Reactivity: Human, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ICFC, IHC, WB; Size: 500 μg
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume.
The purified C11.5 has been tested by blocking fluorochrome conjugated C11.5 for intracellular cytokine staining. In order to obtain complete blocking results, a saturated amount of purified antibody ( 5.0 ug/million cells) should be used for incubation with target cells, prior to staining with fluorochrome conjugated antibody. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: IL-12 p40
Clonality: Monoclonal
Clone: C11.5
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;IL-12/IL-23 p40 subunit (monomer;homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40)
Specifications
- Catalog No:
- 501802-BL
- 501801-BL
- Antigen Symbol:
- IL-12 p40
- IL-12 p40
- Antigen Synonyms:
- IL12 + IL23|TcMF p40|NKSF p40|TSF p40|p40 subunit of IL-12 and IL-23|CLMF p40|Interleukin-12 p40
- IL12 + IL23|TcMF p40|NKSF p40|TSF p40|p40 subunit of IL-12 and IL-23|CLMF p40|Interleukin-12 p40
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- C11.5
- C11.5
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3593
- 3593
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- CHO-expressed, recombinant human IL-12 p70 heterod
- CHO-expressed, recombinant human IL-12 p70 heterod
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Shipping Temperature:
- Ambient RT
- Ambient RT