About this item
Purified anti-FITC [FIT-22]; Isotype: Mouse IgG1, λ; Reactivity: 2nd Step; Apps: FC; Size: 500 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: Fluorescein (FITC)
Clonality: Monoclonal
Clone: FIT-22
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, λ
Reactivity: 2nd Step
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 408301-BL
- 408302-BL
- Antigen Symbol:
- FITC
- FITC
- Antigen Name:
- Fluorescein
- Fluorescein
- Antigen Synonyms:
- FITC
- FITC
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- FIT-22
- FIT-22
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 lambda
- IgG1 lambda
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- FITC conjugated mouse IgG.
- FITC conjugated mouse IgG.
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Shipping Temperature:
- Ambient RT
- Ambient RT