To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:C/EBP b
- Clonality:Monoclonal
- Clone:1H7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- ChIP:Yes
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:12608, 25301
- Antigen Synonyms:Liver-enriched transcriptional activator protein (LAP)|CEBPB|Interleukin 6-dependent DNA-binding protein (IL-6DBP)|Nuclear factor of interleukin 6 (NF-IL6)|Alpha-1-acid glycoprotein/enhancer binding protein|CCAAT/enhancer-binding protein beta
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:Recombinant LIP
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:606202-BL
- Supplier no.:606202
Specifications
About this item
Purified anti-C/EBP β [1H7]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse, Rat; Apps: WB, ChIP, IP, IF; Size: 100 μg
Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 5 μg antibody per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: C/EBP b
Clonality: Monoclonal
Clone: 1H7
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse;Rat