Anti-IgG1, k Isotype Ctrl Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: RTK2071]
Supplier: Biolegend
Certificates
About this item
PerCP/Cy5.5 Rat IgG1, κ Isotype Ctrl [RTK2071]; Isotype: Rat IgG1, κ; Reactivity: Rat; Apps: FC, ICFC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.
Type: Control
Antigen: IgG1, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK2071
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Rat
Specifications
- Catalog No:
- 400426-BL
- 400425-BL
- Antigen Symbol:
- IgG1 k isotype ctrl
- IgG1 k isotype ctrl
- Antibody Type:
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Clone:
- RTK2071
- RTK2071
- Reactivity:
- Rat
- Rat
- Host:
- Rat
- Rat
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Trinitrophenol + KLH
- Trinitrophenol + KLH
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The immunoglobulin was purified by affinity chromatography, and conjugated with PerCP/Cy5.5 under optimal conditions. The solution is free of unconjugated PerCP/Cy5.5 and unconjugated immunoglobulin.
- The immunoglobulin was purified by affinity chromatography, and conjugated with PerCP/Cy5.5 under optimal conditions. The solution is free of unconjugated PerCP/Cy5.5 and unconjugated immunoglobulin.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT