Anti-PSMC2 Rabbit Polyclonal Antibody
Catalog # 89521-160
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Proteasome 26S Subunit, ATPase 2
- Antigen Symbol:PSMC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal
- Gene ID:5701
- Antigen Synonyms:Nbla10058|ATPase 2|S7|macropain) 26S Subunit|MSS1|Proteasome (prosome
- Amino Acid Number:1 to 30
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:48.634 kDa
- Sequence:DYLGADQRKTKEDEKC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This PSMC2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PSMC2.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89521-160
- Supplier no.:AP14584A
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: PSMC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human