Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-CD50 (ICAM-3) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CBR-IC3/1]
  330005-BL
 :  Biolegend
Anti-CD50 (ICAM-3) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CBR-IC3/1]
  330005-BL
 :  Biolegend
 :  330005

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CD50 (ICAM-3)
  • Clonality:
    Monoclonal
  • Clone:
    CBR-IC3/1
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Baboon,
    Cynomolgus monkey,
    Human
  • Size:
    25 tests
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Gene ID:
    3385
  • Antigen Synonyms:
    ICAM-3|ICAM3
  • Storage Temperature:
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
    Ambient RT
  • Immunogen:
    SKW3 cells
  • Purification:
    The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
  • Cat. No.:
    330005-BL
  • Supplier no.:
    330005

 

 

PE anti-human CD50 (ICAM-3) [CBR-IC3/1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Cynomolgus; Apps: FC; Size: 25 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD50 (ICAM-3)
Clonality: Monoclonal
Clone: CBR-IC3/1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Baboon;Cynomolgus