Anti-CPN2 Rabbit Polyclonal Antibody
Catalog # 89512-950
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:carboxypeptidase N, polypeptide 2
- Antigen Symbol:CPN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal
- Gene ID:1370
- Antigen Synonyms:ACBP
- Amino Acid Number:21 to 50
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:60.557 kDa
- Sequence:SDEELATVPLDIPPYC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This CPN2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 21-50 amino acids from the N-terminal region of human CPN2.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89512-950
- Supplier no.:AP13198A
Specifications
About this item
Western Blot: 1:1000, Immunhistochemsitry: 1:10~50,
Type: Primary
Antigen: CPN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: