Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD44 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: IM7]
Anti-CD44 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: IM7]
Supplier:  Biolegend
Certificates
Anti-CD44 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: IM7]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 103019-BL
  • 103020-BL
  • Antigen Symbol:
  • CD44
  • CD44
  • Antigen Synonyms:
  • Hermes|gp85|HUTCH-1|H-CAM|ECMR III|Pgp-1|Ly-24
  • Hermes|gp85|HUTCH-1|H-CAM|ECMR III|Pgp-1|Ly-24
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PB (Pacific Blue)
  • PB (Pacific Blue)
  • Clone:
  • IM7
  • IM7
  • Reactivity:
  • Baboon, Cat, Pig, Horse, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Bovine, Dog, Mouse, Squirrel monkey
  • Baboon, Cat, Pig, Horse, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Bovine, Dog, Mouse, Squirrel monkey
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12505, 960
  • 12505, 960
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Dexamethasone-induced myeloid leukemia M1 cells
  • Dexamethasone-induced myeloid leukemia M1 cells
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™ .
  • The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™ .
  • Concentration:
  • 0.50 mg/mL
  • 0.50 mg/mL
  • Storage Temperature:
  • The CD44 antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The CD44 antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Related Information