Anti-PF4V1 Rabbit Polyclonal Antibody
Catalog # 89515-066
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:platelet factor 4 variant 1
- Antigen Symbol:PF4V1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Gene ID:5197
- Antigen Synonyms:PF4A|SCYB4V1|CXCL4V1|PF4-ALT|CXCL4L1
- Amino Acid Number:77 to 104
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:11.553 kDa
- Sequence:CLLYKKIIKEHLES
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.45mg/ml
- Shipping Temperature:4 °C
- Immunogen:This PF4V1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 77-104 amino acids from the C-terminal region of human PF4V1.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89515-066
- Supplier no.:AP19316B
Specifications
About this item
LEAF (Low Endotoxin, Azide-Free)
Western Blot: 1:1000
Type: Primary
Antigen: PF4V1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human