Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD48 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BJ40]
Anti-CD48 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BJ40]
Supplier:  Biolegend
Certificates
Anti-CD48 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BJ40]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 336705-BL
  • 336706-BL
  • Antigen Symbol:
  • CD48
  • CD48
  • Antigen Synonyms:
  • Blast-1|HuLy-m3
  • Blast-1|HuLy-m3
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • BJ40
  • BJ40
  • Reactivity:
  • Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
  • Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 962
  • 962
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
  • The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Related Information