Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ANKDD1B Rabbit Polyclonal Antibody
Anti-ANKDD1B Rabbit Polyclonal Antibody
Catalog # 89515-792
Supplier:  Abgent
Anti-ANKDD1B Rabbit Polyclonal Antibody
Catalog # 89515-792
Supplier:  Abgent
Supplier Number:  AP18760B

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    ANKDD1B
  • Antigen Symbol:
    ANKDD1B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Epitope:
    C-Terminal
  • Gene ID:
    728780
  • Amino Acid Number:
    494 to 520
  • Storage Buffer:
    PBS with 0.09% (W/V) sodium azide, pH7.4
  • Molecular Weight:
    59.088 kDa
  • Sequence:
    EKTRHFKSKTDSC
  • Storage Temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This ANKDD1B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 494-520 amino acids from the C-terminal region of human ANKDD1B.
  • Purification:
    Protein A & Peptide affinity purification
  • Cat. No.:
    89515-792
  • Supplier no.:
    AP18760B

Specifications

About this item

Western Blot: 1:1000

Type: Primary
Antigen: ANKDD1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human