- Description:LYFO DISC® Escherichia coli (serotype O157:H7)
- Storage Conditions:Easy storage; no dry ice required; reduces shipping costs
- Traceability:Assured Quality - Traceable to reference culture
- Cat. No.:89505-270
- ATCC no.:ATCC® 35150™*
- Supplier no.:0617L
LYFO DISK® is a lyophilized pellet containing a single strain of a QC microorganism and is the most economical reference stock culture option
LYFO DISK® is known for its ease-of-use; simply re-hydrate the pellet and inoculate. LYFO DISK® is available in a re-sealable vial containing 6 pellets of a single QC microorganism strain.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.