Order Entry
Puerto Rico
ContactUsLinkComponent
CT-2000 Series Constant Temperature Bath, 240 V, Cannon Instrument Company
CT-2000 Series Constant Temperature Bath, 240 V, Cannon Instrument Company
Catalog # 47735-076
Supplier:  Cannon
Irregular Voltage
Small Business Enterprise
undefined
CT-2000 Series Constant Temperature Bath, 240 V, Cannon Instrument Company
Catalog # 47735-076
Supplier:  Cannon
Supplier Number:  9726-A35
Irregular Voltage
Small Business Enterprise

Specifications

  • Description:
    CT-2000F Constant Temperature Bath
  • Electrical:
    240V, 220-240VAC, 50/60Hz
  • Cat. No.:
    47735-076
  • Supplier no.:
    9726-A35

Specifications

About this item

Convenient keypad operation and ASTM reliability at temperatures to 150°C make the CT-2000 the bath of choice for many users

  • 240 V model, not for use in USA

Microprocessor-controlled temperature regulation, on-screen temperature monitoring, and serial networking options make this digital bath an attractive and fully functional addition to the modern laboratory.

Caution: 240 V model, not for use in USA. Corresponding model with voltage suitable for use in US (47735-074) is also available, please see separate product page.