Isoflurane USP Reference Standard
1 ml
Catalog # 101095-254
Marketsource
Supplier: USP
: 1349003
Isoflurane USP Reference Standard
Catalog # 101095-254
Supplier: USP
Marketsource
CAS Number: 26675-46-7
Jump to:
Product Details & Documents
Synonyms: 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether
- CAS Number:26675-46-7
- Formula:C₃H₂ClF₅O
- MDL Number:MFCD00066609
- Molecular Weight:184.49 g/mol
- Boiling Pt:47…50 °C
- EINECS:247-897-7
- UN:3334
- ADR:9,III
Specifications
Specification Test Results
- Molecular formula:C3H2ClF5O
- Molecular weight:184.49
- Volatile:100 %
- Specific gravity:1.45 - 1.496
- Synonym:1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether
Specifications
- Size:1 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



