Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Acetone ≥99.5%, Technical Grade

Acetone ≥99.5%, Technical Grade

Flammable
Irritant
Marketsource
Supplier:   Aqua Solutions
Acetone ≥99.5%, Technical Grade
Supplier:  Aqua Solutions
Marketsource
Certificates
Acetone ≥99.5%, Technical Grade

1 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • UN:1090
  • ADR:3,II

Documents

Specifications for Product Family

Specification Test Results

  • Assay:
    99.5% Min.
  • Color (APHA):
    Max. 10
  • Solubility in Water:
    Passes Test
  • Aldehyde (As HCHO):
    Max. 0.002%
  • Isopropyl alcohol:
    Max. 0.05 %
  • Evaporation residue:
    Max. 0.001%
  • Titrable acid:
    Max. 0.0003 meq/g
  • Titrable base:
    Max. 0.0006 meq/g
  • Water:
    Max. 0.5 %
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle, amber
    500 ml
    Discontinued
    Supplier Number: A1501-500ML
    Specifications:
    Marketsource
    Marketsource
  • Metal oblong can
    4 L
    Discontinued
    Supplier Number: A1501-4L
    Specifications:
    Marketsource
    Marketsource
  • Reike can, black
    20 L
    Discontinued
    Supplier Number: A1501-20L
    Specifications:
    Marketsource
    Marketsource
  • Metal drum
    200 L
    Log in for availability
    Log in for availability
    Each (200l)
    Each (200l) - $2,152.29
    $2,152.29
    Each (200l)
       $2,152.29

    Supplier Number: A1501-200L
    Specifications:
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.