Orders LinkContactUsLinkComponent
Solid Brass Mass Set
Solid Brass Mass Set
Solid Brass Mass Set
Catalog # 470206-078
Supplier:   Wards
Supplier #:  PH0037H
Solid Brass Mass Set
Catalog # 470206-078
Supplier:  Wards
Solid Brass Mass Set
Solid Brass Mass Set
Solid Brass Mass Set
Clearance
$51.70
Sale
Valid until 31/12/2026
Log in for availability
Each
Product Details & Documents
Solid Brass. Accurate machined masses placed on a plastic and metal rack with handle.
Set of 12; 1 x 1g, 2 x 2g, 1 x 5g, 1 x 10g, 2 x 20g, 1 x 50g, 1 x 100g, 2 x 200g, 1 x 500g. Total 1110g.
Specifications
  • Description:
    Solid Brass Mass Set