ContactUsLinkComponent
Utility Jar
Utility Jar
Catalog # 470173-740
Supplier:  Dynalon
undefined
Utility Jar
Catalog # 470173-740
Supplier:  Dynalon
Supplier Number:  600335
Specifications
  • Description:
    Utility Jar
  • Cat. No.:
    470173-740
  • Supplier no.:
    600335
Specifications
About this item
One gallon capacity, high density polyethylene utility jar with metal closure.
  • Wide mouth, natural color
  • Withstands temperatures to 110 °C
Hundreds of uses around the lab.