BS/U-Tube Viscometer, Cannon Instrument
Supplier: Cannon
Certificates
About this item
The BS/U-Tube Viscometer measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104
Viscometer is calibrated. Minimum sample volume 7 mL in sizes A to C, 12 mL in sizes D to F, and 23 mL in sizes G and H. Minimum bath depth of 11".
Frequently Bought Together